3,8-Dioxo-2,3,5,6,7,8-hexahydro-4-isoquinolinecarboxamide ≥95%

Supplier: Matrix Scientific

045707-500MG 045707-5G 045707-1G
101872-208EA 135.7 USD
101872-208 101872-212 101872-210
3,8-Dioxo-2,3,5,6,7,8-hexahydro-4-isoquinolinecarboxamide ≥95%
3,8-Dioxo-2,3,5,6,7,8-hexahydro-4-isoquinolinecarboxamide

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₃
Melting Pt: 315…317 °C
MDL Number: MFCD03425819
CAS Number: 171113-31-8

Order Now

Specification Test Results

Melting point 315-317°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR