5-Methoxy-2-(1H-pyrrol-1-yl)aniline ≥95%
Supplier: Matrix Scientific
045697-1G
045697-5G
045697-500MG
101872-160EA
153.85
USD
101872-160
101872-164
101872-162
5-Methoxy-2-(1H-pyrrol-1-yl)aniline ≥95%
5-Methoxy-2-(1H-pyrrol-1-yl)aniline
Hazard: Irritant
Formula:
C₁₁H₁₂N₂O Melting Pt: 41…43 °C |
MDL Number:
MFCD00047063 CAS Number: 59194-26-2 |
Specification Test Results
Melting point | 41-43°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...