Methyl-3-isothiocyanato-2-thiophenecarboxylate ≥95%

Supplier: Matrix Scientific

045686-1G 045686-5G 045686-500MG
101872-106EA 2670.77 USD
101872-106 101872-104 101872-102
Methyl-3-isothiocyanato-2-thiophenecarboxylate ≥95%
Methyl-3-isothiocyanato-2-thiophenecarboxylate

Hazard: Irritant

Formula: C₇H₅NO₂S₂
Melting Pt: 57…58 °C
MDL Number: MFCD00052589
CAS Number: 81321-10-0

Order Now

Specification Test Results

Melting point 57-58°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR