2-Oxo-4-(2-thienyl)-1,2-dihydro-3-pyridinecarbonitrile ≥95%
Supplier: Matrix Scientific
045669-5G
045669-500MG
045669-1G
101872-020EA
375.38
USD
101872-020
101872-024
101872-022
2-Oxo-4-(2-thienyl)-1,2-dihydro-3-pyridinecarbonitrile ≥95%
2-Oxo-4-(2-thienyl)-1,2-dihydro-3-pyridinecarbonitrile
Hazard: Irritant
Formula:
C₁₀H₆N₂OS Melting Pt: 291…293 °C |
MDL Number:
MFCD03001295 CAS Number: 174713-67-8 |
Specification Test Results
Melting point | 291-293°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...