6-Bromo-1,3-benzodioxole-5-carbonitrile ≥95%
Supplier: Matrix Scientific
045659-1G
045659-500MG
101871-974EA
556.92
USD
101871-974
101871-976
6-Bromo-1,3-benzodioxole-5-carbonitrile ≥95%
6-Bromo-1,3-benzodioxole-5-carbonitrile
Hazard: Irritant
Formula:
C₈H₄BrNO₂ MW: 226.04 g/mol Melting Pt: 104…106 °C |
MDL Number:
MFCD02571372 CAS Number: 6120-26-9 |
Specification Test Results
Melting point | 104-106°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...