4-(4-Bromophenyl)-2-hydrazino-1,3-thiazole ≥95%

Supplier: Matrix Scientific

045657-500MG 045657-1G 045657-5G
101871-966EA 467.69 USD
101871-966 101871-968 101871-964
4-(4-Bromophenyl)-2-hydrazino-1,3-thiazole ≥95%
4-(4-Bromophenyl)-2-hydrazino-1,3-thiazole

Hazard: Irritant

Formula: C₉H₈BrN₃S
Melting Pt: 180…182 °C (dec.)
MDL Number: MFCD01445245
CAS Number: 4871-22-1

Order Now

Specification Test Results

Melting point 180-182°C(d

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR