5-Bromo-2-(2,5-dimethyl-1H-pyrrol-1-yl)pyrimidine ≥95%

Supplier: Matrix Scientific

045653-5G 045653-1G 045653-500MG
101871-944EA 350.77 USD
101871-944 101871-946 101871-942
5-Bromo-2-(2,5-dimethyl-1H-pyrrol-1-yl)pyrimidine ≥95%
5-Bromo-2-(2,5-dimethyl-1H-pyrrol-1-yl)pyrimidine

Hazard: Irritant

Formula: C₁₀H₁₀BrN₃
Melting Pt: 66…68 °C
MDL Number: MFCD02082433
CAS Number: 478258-81-0

Order Now

Specification Test Results

Melting point 66-68°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR