1-Methyl-N-[3-(trifluoromethyl)phenyl]-1-hydrazinecarbothioamide ≥95%
Supplier: Matrix Scientific
045636-5G
045636-1G
045636-500MG
101871-868EA
278.46
USD
101871-868
101871-870
101871-872
1-Methyl-N-[3-(trifluoromethyl)phenyl]-1-hydrazinecarbothioamide ≥95%
1-Methyl-N-[3-(trifluoromethyl)phenyl]-1-hydrazinecarbothioamide
Hazard: Irritant
Formula:
C₉H₁₀F₃N₃S Melting Pt: 130…132 °C |
MDL Number:
MFCD08056673 CAS Number: 70483-65-7 |
Specification Test Results
Melting point | 130-132°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...