1-[3-(2-Pyrimidinyloxy)phenyl]-1-ethanone ≥95%
Supplier: Matrix Scientific
045632-500MG
045632-5G
045632-1G
101871-846EA
192.31
USD
101871-846
101871-848
101871-844
1-[3-(2-Pyrimidinyloxy)phenyl]-1-ethanone ≥95%
1-[3-(2-Pyrimidinyloxy)phenyl]-1-ethanone
Hazard: Irritant
Formula:
C₁₂H₁₀N₂O₂ MW: 214.23 g/mol Melting Pt: 84…86 °C |
MDL Number:
MFCD01568700 CAS Number: 339105-37-2 |
Specification Test Results
Melting point | 84-86°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...