5-(3-Oxo-3-phenyl-1-propenyl)-2-furaldehyde ≥95%
Supplier: Matrix Scientific
045625-1G
045625-500MG
045625-5G
101871-804EA
921.54
USD
101871-804
101871-806
101871-808
5-(3-Oxo-3-phenyl-1-propenyl)-2-furaldehyde ≥95%
5-(3-Oxo-3-phenyl-1-propenyl)-2-furaldehyde
Hazard: Irritant
Formula:
C₁₄H₁₀O₃ MW: 226.23 g/mol Melting Pt: 105…107 °C |
MDL Number:
MFCD01315261 CAS Number: 6575-75-3 |
Specification Test Results
Melting point | 105-107°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...