3-Chloro-N-(2-chloro-3-pyridinyl)-2,2-dimethylpropanamide ≥95%
Supplier: Matrix Scientific
045611-500MG
045611-5G
045611-1G
101871-742EA
467.69
USD
101871-742
101871-744
101871-740
3-Chloro-N-(2-chloro-3-pyridinyl)-2,2-dimethylpropanamide ≥95%
3-Chloro-N-(2-chloro-3-pyridinyl)-2,2-dimethylpropanamide
Hazard: Irritant
Formula:
C₁₀H₁₂Cl₂N₂O MW: 247.13 g/mol Melting Pt: 66…69 °C |
MDL Number:
MFCD00793432 CAS Number: 339100-93-5 |
Specification Test Results
Melting point | 66-69°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...