5-Chloro-1-methyl-3-phenyl-1H-pyrazole-4-carbonitrile ≥95%
Supplier: Matrix Scientific
045607-500MG
045607-5G
045607-1G
101871-720EA
698.46
USD
101871-720
101871-722
101871-718
5-Chloro-1-methyl-3-phenyl-1H-pyrazole-4-carbonitrile ≥95%
5-Chloro-1-methyl-3-phenyl-1H-pyrazole-4-carbonitrile
Hazard: Irritant
Formula:
C₁₁H₈ClN₃ Melting Pt: 101…105 °C |
MDL Number:
MFCD01935950 CAS Number: 321848-49-1 |
Specification Test Results
Melting point | 101-105°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...