2-[(4-Chlorobenzyl)sulfanyl]benzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

045595-500MG 045595-5G 045595-1G
101871-666EA 350.77 USD
101871-666 101871-668 101871-664
2-[(4-Chlorobenzyl)sulfanyl]benzenecarboxylic acid ≥95%
2-[(4-Chlorobenzyl)sulfanyl]benzenecarboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₁ClO₂S
Melting Pt: 212…215 °C
MDL Number: MFCD01315364
CAS Number: 15887-84-0

Order Now

Specification Test Results

Melting point 212-215°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR