4-[(2-Aminoethyl)amino]benzenecarboxylic acid dihydrochloride ≥97%
Supplier: Matrix Scientific
045592-500MG
045592-1G
045592-5G
101871-654EA
350.77
USD
101871-654
101871-656
101871-652
4-[(2-Aminoethyl)amino]benzenecarboxylic acid dihydrochloride ≥97%
4-[(2-Aminoethyl)amino]benzenecarboxylic acid dihydrochloride
Hazard: Irritant
Formula:
C₉H₁₄Cl₂N₂O₂ Melting Pt: 289 °C |
MDL Number:
MFCD00664237 CAS Number: 148720-10-9 |
Specification Test Results
Melting point | 289°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...