6-Chloro-2H-chromene-3-carbonitrile ≥95%
Supplier: Matrix Scientific
044420-1G
044420-500MG
101865-744EA
350.77
USD
101865-744
101865-742
6-Chloro-2H-chromene-3-carbonitrile ≥95%
6-Chloro-2H-chromene-3-carbonitrile
Hazard: Irritant
Formula:
C₁₀H₆ClNO Melting Pt: 135…137 °C |
MDL Number:
MFCD00052359 CAS Number: 57543-67-6 |
Specification Test Results
Melting point | 135-137°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...