Methyl-2H-chromene-3-carboxylate ≥95%

Supplier: Matrix Scientific

044416-1G 044416-500MG 044416-5G
101865-722EA 927.69 USD
101865-722 101865-720 101865-724
Methyl-2H-chromene-3-carboxylate ≥95%
Methyl-2H-chromene-3-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₀O₃
MW: 190.2 g/mol
Melting Pt: 55…57 °C
MDL Number: MFCD00202022
CAS Number: 36044-49-2

Order Now

Specification Test Results

Melting point 55-57°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR