[1-Methyl-5-(phenylsulfanyl)-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol ≥95%
Supplier: Matrix Scientific
Synonyms:
1-Methyl-5-(phenylthio)-3-(trifluoromethyl)-1H-pyrazol-4-ylmethanol
044415-1G
044415-500MG
044415-5G
101865-718EA
2129.23
USD
101865-718
101865-716
101865-714
[1-Methyl-5-(phenylsulfanyl)-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol ≥95%
[1-Methyl-5-(phenylsulfanyl)-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol
Hazard: Irritant
Formula:
C₁₂H₁₁F₃N₂OS MW: 288.29 g/mol Melting Pt: 45…46 °C |
MDL Number:
MFCD00172634 CAS Number: 318239-51-9 |
Specification Test Results
Melting point | 45-46°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...