(1-Phenyl-1H-1,2,3-triazol-4-yl)methanol ≥95%
Supplier: Matrix Scientific
044400-500MG
044400-5G
044400-1G
101865-644EA
323.08
USD
101865-644
101865-642
101865-640
(1-Phenyl-1H-1,2,3-triazol-4-yl)methanol ≥95%
(1-Phenyl-1H-1,2,3-triazol-4-yl)methanol
Hazard: Irritant
Formula:
C₉H₉N₃O MW: 175.1900 g/mol Melting Pt: 114…116 °C |
MDL Number:
MFCD00100214 CAS Number: 103755-58-4 |
Specification Test Results
Melting point | 114-116°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...