5-Amino-1-(4-methoxyphenyl)-1H-pyrazole-4-carbonitrile ≥95%

Supplier: Matrix Scientific

044362-5G 044362-500MG 044362-1G
101865-478EA 350.77 USD
101865-478 101865-476 101865-480
5-Amino-1-(4-methoxyphenyl)-1H-pyrazole-4-carbonitrile ≥95%
5-Amino-1-(4-methoxyphenyl)-1H-pyrazole-4-carbonitrile

Hazard: Irritant

Formula: C₁₁H₁₀N₄O
Melting Pt: 125…128 °C
MDL Number: MFCD00128343
CAS Number: 116884-64-1

Order Now

Specification Test Results

Melting point 125-128°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR