2-Propyl-1,3-benzoxazol-6-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Amino-2-propyl-1,3-benzoxazole

044360-500MG 044360-1G 044360-5G
101865-468EA 1600 USD
101865-468 101865-466 101865-464
2-Propyl-1,3-benzoxazol-6-amine ≥95%
2-Propyl-1,3-benzoxazol-6-amine

Hazard: Irritant

Formula: C₁₀H₁₂N₂O
MW: 176.22 g/mol
Melting Pt: 83…85 °C
MDL Number: MFCD06200963
CAS Number: 875851-66-4

Order Now

Specification Test Results

Melting point 83-85°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR