Methyl-3-iodoimidazo[1,2-a]pyridine-6-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 3-iodoimidazo[1,2-a]pyridine-6-carboxylate
044358-500MG
044358-1G
044358-5G
101865-456EA
2670.77
USD
101865-456
101865-454
101865-452
Methyl-3-iodoimidazo[1,2-a]pyridine-6-carboxylate ≥95%
Methyl 3-iodoimidazo[1,2-a]pyridine-6-carboxylate
Hazard: Irritant
Formula:
C₉H₇IN₂O₂ MW: 302.0686 g/mol Melting Pt: 188…191 °C |
MDL Number:
MFCD07021385 CAS Number: 460087-82-5 |
Specification Test Results
Melting point | 188-191°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...