3-(1,4-Dioxaspiro[4.5]dec-7-en-8-yl)-1H-indole ≥95%
Supplier: Matrix Scientific
044338-5G
044338-500MG
044338-1G
101865-354EA
397.9
USD
101865-354
101865-352
101865-350
3-(1,4-Dioxaspiro[4.5]dec-7-en-8-yl)-1H-indole ≥95%
3-(1,4-Dioxaspiro[4.5]dec-7-en-8-yl)-1H-indole
Hazard: Irritant
Formula:
C₁₆H₁₇NO₂ Melting Pt: 194…196 °C |
MDL Number:
MFCD06411611 CAS Number: 143165-32-6 |
Specification Test Results
Melting point | 194-196°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...