2-(1,4,5,6-Tetrahydro-2-pyrimidinyl)thiophenol ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(1,4,5,6-Tetrahydropyrimidin-2-yl)benzenethiol

044334-5G 044334-1G 044334-500MG
101865-342EA 1461.54 USD
101865-342 101865-340 101865-338
2-(1,4,5,6-Tetrahydro-2-pyrimidinyl)thiophenol ≥95%
2-(1,4,5,6-Tetrahydro-2-pyrimidinyl)thiophenol

Hazard: Irritant

Formula: C₁₀H₁₂N₂S
Melting Pt: 235…237 °C
MDL Number: MFCD00052612
CAS Number: 53440-32-7

Order Now

Specification Test Results

Melting point 235-237°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR