4-Morpholino-3-nitrobenzaldehyde ≥95%
Supplier: Matrix Scientific
044311-500MG
044311-1G
044311-5G
101865-226EA
806.15
USD
101865-226
101865-224
101865-222
4-Morpholino-3-nitrobenzaldehyde ≥95%
4-Morpholino-3-nitrobenzaldehyde
Hazard: Irritant
Formula:
C₁₁H₁₂N₂O₄ Melting Pt: 57…59 °C |
CAS Number:
300541-91-7 |
Specification Test Results
Melting point | 57-59°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...