Methyl-2-(4-nitrophenoxy)benzenecarboxylate ≥95%

Supplier: Matrix Scientific

044310-1G 044310-500MG 044310-5G
101865-216EA 181.54 USD
101865-216 101865-220 101865-218
Methyl-2-(4-nitrophenoxy)benzenecarboxylate ≥95%
Methyl-2-(4-nitrophenoxy)benzenecarboxylate

Hazard: Irritant

Formula: C₁₄H₁₁NO₅
Melting Pt: 74…77 °C
MDL Number: MFCD04117807
CAS Number: 212189-50-9

Order Now

Specification Test Results

Melting point 74-77°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR