(2,5-Dichloro-3-pyridinyl)methanol ≥95%
Supplier: Matrix Scientific
Synonyms:
2,5-Dichloropyridine-3-methanol
044309-5G
044309-500MG
044309-1G
101865-214EA
480
USD
101865-214
101865-212
101865-210
(2,5-Dichloro-3-pyridinyl)methanol ≥95%
(2,5-Dichloro-3-pyridinyl)methanol
Hazard: Irritant
Formula:
C₆H₅Cl₂NO MW: 178.02 g/mol Melting Pt: 64…66 °C |
MDL Number:
MFCD06659487 CAS Number: 558465-93-3 |
Specification Test Results
Melting point | 64-66°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...