5-Bromo-2-chloro-4,6-dimethylnicotinonitrile ≥95%
Supplier: Matrix Scientific
044305-1G
044305-500MG
044305-5G
101865-196EA
80.5
USD
101865-196
101865-194
101865-192
5-Bromo-2-chloro-4,6-dimethylnicotinonitrile ≥95%
5-Bromo-2-chloro-4,6-dimethylnicotinonitrile
Hazard: Irritant
Formula:
C₈H₆BrClN₂ Melting Pt: 106…108 °C |
MDL Number:
MFCD03791216 CAS Number: 42951-71-3 |
Specification Test Results
Melting point | 106-108°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...