5-(5-Bromo-2-thienyl)-1H-pyrazole ≥95%
Supplier: Matrix Scientific
Synonyms:
5-(5-Bromothiophen-2-yl)-1H-pyrazole
044300-5G
044300-1G
044300-500MG
101865-168EA
458.46
USD
101865-168
101865-166
101865-164
5-(5-Bromo-2-thienyl)-1H-pyrazole ≥95%
5-(5-Bromo-2-thienyl)-1H-pyrazole
Hazard: Irritant
Formula:
C₇H₅BrN₂S MW: 229.1 g/mol Melting Pt: 167…168 °C |
MDL Number:
MFCD00120516 CAS Number: 166196-67-4 |
Specification Test Results
Melting point | 167-168°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...