1-(5-Chloro-2-nitrophenyl)-1-ethanone ≥95%
Supplier: Matrix Scientific
Synonyms:
5'-Chloro-2'-nitroacetophenone
, 1-(5-Chloro-2-nitrophenyl)ethanone
044292-1G
044292-5G
101865-132EA
272.31
USD
101865-132
101865-130
1-(5-Chloro-2-nitrophenyl)-1-ethanone ≥95%
1-(5-Chloro-2-nitrophenyl)-1-ethanone
Hazard: Irritant
Formula:
C₈H₆ClNO₃ MW: 199.59 g/mol Melting Pt: 58…61 °C |
MDL Number:
MFCD03787322 CAS Number: 18640-60-3 |
Specification Test Results
Melting point | 58-61°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...