3-(4-Chlorophenyl)-3-[(2,2,2-trifluoroacetyl)amino]propanamide ≥95%
Supplier: Matrix Scientific
044286-5G
044286-500MG
044286-1G
101865-106EA
556.92
USD
101865-106
101865-110
101865-108
3-(4-Chlorophenyl)-3-[(2,2,2-trifluoroacetyl)amino]propanamide ≥95%
3-(4-Chlorophenyl)-3-[(2,2,2-trifluoroacetyl)amino]propanamide
Hazard: Irritant
Formula:
C₁₁H₁₀ClF₃N₂O₂ Melting Pt: 241…243 °C |
MDL Number:
MFCD03012828 CAS Number: 439110-82-4 |
Specification Test Results
Melting point | 241-243°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...