4-(3-Bromo-2-thienyl)-2-pyrimidinamine ≥95%
Supplier: Matrix Scientific
044260-500MG
101864-988EA
556.92
USD
101864-988
4-(3-Bromo-2-thienyl)-2-pyrimidinamine ≥95%
4-(3-Bromo-2-thienyl)-2-pyrimidinamine
Hazard: Irritant
Formula:
C₈H₆BrN₃S Melting Pt: 146…147 °C |
MDL Number:
MFCD03791202 CAS Number: 886360-54-9 |
Specification Test Results
Melting point | 146-147°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...