1-(3-Bromo-2-thienyl)-3-(dimethylamino)-2-propen-1-one ≥95%

Supplier: Matrix Scientific

044259-5G 044259-500MG 044259-1G
101864-982EA 375.38 USD
101864-982 101864-984 101864-986
1-(3-Bromo-2-thienyl)-3-(dimethylamino)-2-propen-1-one ≥95%
1-(3-Bromo-2-thienyl)-3-(dimethylamino)-2-propen-1-one

Hazard: Irritant

Formula: C₉H₁₀BrNOS
Melting Pt: 63…64 °C
MDL Number: MFCD03618050
CAS Number: 166196-82-3

Order Now

Specification Test Results

Melting point 63-64°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR