6-(4-Bromophenyl)imidazo[2,1-b][1,3]thiazole-5-carbaldehyde ≥95%

Supplier: Matrix Scientific

044244-500MG 044244-5G 044244-1G
101864-912EA 806.15 USD
101864-912 101864-908 101864-910
6-(4-Bromophenyl)imidazo[2,1-b][1,3]thiazole-5-carbaldehyde ≥95%
6-(4-Bromophenyl)imidazo[2,1-b][1,3]thiazole-5-carbaldehyde

Hazard: Irritant

Formula: C₁₂H₇BrN₂OS
Melting Pt: 188…190 °C
MDL Number: MFCD00268738
CAS Number: 451485-66-8

Order Now

Specification Test Results

Melting point 188-190°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR