1-(4-Nitrobenzoyl)tetrahydro-4(1H)-pyridinone ≥95%
Supplier: Matrix Scientific
044229-1G
044229-500MG
044229-5G
101864-838EA
278.46
USD
101864-838
101864-840
101864-842
1-(4-Nitrobenzoyl)tetrahydro-4(1H)-pyridinone ≥95%
1-(4-Nitrobenzoyl)tetrahydro-4(1H)-pyridinone
Hazard: Irritant
Formula:
C₁₂H₁₂N₂O₄ Melting Pt: 118…120 °C |
MDL Number:
MFCD03012121 CAS Number: 34259-84-2 |
Specification Test Results
Melting point | 118-120°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...