7-(Benzyloxy)-2-naphthol ≥95%

Supplier: Matrix Scientific

044227-500MG 044227-1G
101864-830EA 278.46 USD
101864-830 101864-832
7-(Benzyloxy)-2-naphthol ≥95%
7-(Benzyloxy)-2-naphthol

Hazard: Irritant

Formula: C₁₇H₁₄O₂
Melting Pt: 151…152 °C
MDL Number: MFCD03425751
CAS Number: 118495-07-1

Order Now

Specification Test Results

Melting point 151-152°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR