Methyl-1-[(4-methoxyphenyl)sulfonyl]-4-piperidinecarboxylate ≥95%

Supplier: Matrix Scientific

044225-5G 044225-500MG 044225-1G
101864-818EA 278.46 USD
101864-818 101864-820 101864-822
Methyl-1-[(4-methoxyphenyl)sulfonyl]-4-piperidinecarboxylate ≥95%
Methyl-1-[(4-methoxyphenyl)sulfonyl]-4-piperidinecarboxylate

Hazard: Irritant

Formula: C₁₄H₁₉NO₅S
Melting Pt: 131…133 °C
MDL Number: MFCD02736078
CAS Number: 349622-40-8

Order Now

Specification Test Results

Melting point 131-133°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR