Ethyl-6-iodoimidazo[1,2-a]pyridine-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 6-iodoimidazo[1,2-a]pyridine-2-carboxylate

044207-1G 044207-500MG 044207-5G
101864-724EA 467.69 USD
101864-724 101864-726 101864-722
Ethyl-6-iodoimidazo[1,2-a]pyridine-2-carboxylate ≥95%
Ethyl 6-iodoimidazo[1,2-a]pyridine-2-carboxylate

Hazard: Irritant

Formula: C₁₀H₉IN₂O₂
MW: 316.1000 g/mol
Melting Pt: 177…179 °C
MDL Number: MFCD02186151
CAS Number: 214958-32-4

Order Now

Specification Test Results

Melting point 177-179°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR