4H-1,2,4-Triazole-3,4,5-triamine ≥95%

Supplier: Matrix Scientific

044200-500MG
101864-684EA 208.15 USD
101864-684
4H-1,2,4-Triazole-3,4,5-triamine ≥95%
4H-1,2,4-Triazole-3,4,5-triamine

Hazard: Irritant

Formula: C₂H₆N₆
Melting Pt: 269…271 °C
MDL Number: MFCD00628183
CAS Number: 473-96-1

Order Now

Specification Test Results

Melting point 269-271°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR