6-Cyclopropyl-2-oxo-4-(trifluoromethyl)-1,2-dihydro-3-pyridinecarbonitrile ≥95%

Supplier: Matrix Scientific

044183-500MG 044183-1G
101864-592EA 292.31 USD
101864-592 101864-594
6-Cyclopropyl-2-oxo-4-(trifluoromethyl)-1,2-dihydro-3-pyridinecarbonitrile ≥95%
6-Cyclopropyl-2-oxo-4-(trifluoromethyl)-1,2-dihydro-3-pyridinecarbonitrile

Hazard: Irritant

Formula: C₁₀H₇F₃N₂O
MW: 228.18 g/mol
Melting Pt: 229…232 °C
MDL Number: MFCD04125115
CAS Number: 861212-38-6

Order Now

Specification Test Results

Melting point 229-232°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR