2-(2,4-Difluorophenoxy)-1-methyl-1H-indole-3-carbaldehyde ≥95%
Supplier: Matrix Scientific
044178-5G
044178-1G
044178-500MG
101864-564EA
747.69
USD
101864-564
101864-566
101864-568
2-(2,4-Difluorophenoxy)-1-methyl-1H-indole-3-carbaldehyde ≥95%
2-(2,4-Difluorophenoxy)-1-methyl-1H-indole-3-carbaldehyde
Hazard: Irritant
Formula:
C₁₆H₁₁F₂NO₂ Melting Pt: 138…140 °C |
MDL Number:
MFCD01568349 CAS Number: 338416-86-7 |
Specification Test Results
Melting point | 138-140°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...