2-(2,3-Dimethylphenoxy)-1-methyl-1H-indole-3-carbaldehyde ≥95%
Supplier: Matrix Scientific
044176-1G
044176-500MG
044176-5G
101864-552EA
747.69
USD
101864-552
101864-554
101864-556
2-(2,3-Dimethylphenoxy)-1-methyl-1H-indole-3-carbaldehyde ≥95%
2-(2,3-Dimethylphenoxy)-1-methyl-1H-indole-3-carbaldehyde
Hazard: Irritant
Formula:
C₁₈H₁₇NO₂ Melting Pt: 148…150 °C |
MDL Number:
MFCD01315834 CAS Number: 338416-82-3 |
Specification Test Results
Melting point | 148-150°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...