1-Methyl-2-phenoxy-1H-indole-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

044175-5G 044175-500MG 044175-1G
101864-548EA 927.69 USD
101864-548 101864-550 101864-546
1-Methyl-2-phenoxy-1H-indole-3-carbaldehyde ≥95%
1-Methyl-2-phenoxy-1H-indole-3-carbaldehyde

Hazard: Irritant

Formula: C₁₆H₁₃NO₂
Melting Pt: 122…124 °C
MDL Number: MFCD01315833
CAS Number: 338416-80-1

Order Now

Specification Test Results

Melting point 122-124°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR