2-[(4-Chlorobenzyl)sulfanyl]-1-methyl-1H-indole-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

044170-500MG 044170-1G 044170-5G
101864-516EA 921.54 USD
101864-516 101864-518 101864-520
2-[(4-Chlorobenzyl)sulfanyl]-1-methyl-1H-indole-3-carbaldehyde ≥95%
2-[(4-Chlorobenzyl)sulfanyl]-1-methyl-1H-indole-3-carbaldehyde

Hazard: Irritant

Formula: C₁₇H₁₄ClNOS
Melting Pt: 123…126 °C
MDL Number: MFCD01315823
CAS Number: 338416-58-3

Order Now

Specification Test Results

Melting point 123-126°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR