1-Methyl-2-(4-methylphenoxy)-1H-indole-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

044167-5G 044167-1G 044167-500MG
101864-498EA 747.69 USD
101864-498 101864-500 101864-502
1-Methyl-2-(4-methylphenoxy)-1H-indole-3-carbaldehyde ≥95%
1-Methyl-2-(4-methylphenoxy)-1H-indole-3-carbaldehyde

Hazard: Irritant

Formula: C₁₇H₁₅NO₂
Melting Pt: 126…128 °C
MDL Number: MFCD01315813
CAS Number: 338416-21-0

Order Now

Specification Test Results

Melting point 126-128°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR