8-Amino-3-(3,4-dichlorophenyl)imidazo[1,2-a]pyridine ≥95%
Supplier: Matrix Scientific
044159-5G
044159-1G
044159-500MG
101864-460EA
2129.23
USD
101864-460
101864-456
101864-458
8-Amino-3-(3,4-dichlorophenyl)imidazo[1,2-a]pyridine ≥95%
8-Amino-3-(3,4-dichlorophenyl)imidazo[1,2-a]pyridine
Hazard: Irritant
Formula:
C₁₃H₉Cl₂N₃ Melting Pt: 173…175 °C |
MDL Number:
MFCD00975027 CAS Number: 338415-23-9 |
Specification Test Results
Melting point | 173-175°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...