2-Methyl-1-(4-methylbenzyl)-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarbonitrile ≥95%
Supplier: Matrix Scientific
044158-1G
044158-500MG
044158-5G
101864-450EA
375.38
USD
101864-450
101864-452
101864-454
2-Methyl-1-(4-methylbenzyl)-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarbonitrile ≥95%
2-Methyl-1-(4-methylbenzyl)-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarbonitrile
Hazard: Irritant
Formula:
C₁₅H₁₆N₂O MW: 240.31 g/mol Melting Pt: 105…107 °C |
MDL Number:
MFCD00975017 CAS Number: 338415-13-7 |
Specification Test Results
Melting point | 105-107°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...