1-(2,6-Dichlorobenzyl)-1H-indole-2,3-dione ≥95%
Supplier: Matrix Scientific
044151-5G
044151-500MG
044151-1G
101864-420EA
181.54
USD
101864-420
101864-422
101864-424
1-(2,6-Dichlorobenzyl)-1H-indole-2,3-dione ≥95%
1-(2,6-Dichlorobenzyl)-1H-indole-2,3-dione
Hazard: Irritant
Formula:
C₁₅H₉Cl₂NO₂ Melting Pt: 197…199 °C |
MDL Number:
MFCD00141590 CAS Number: 303997-00-4 |
Specification Test Results
Melting point | 197-199°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...