5-Acetyl-1-(2-chloro-3-pyridinyl)-2,4(1H,3H)-pyrimidinedione ≥95%

Supplier: Matrix Scientific

044148-1G 044148-5G 044148-500MG
101864-406EA 693.45 USD
101864-406 101864-408 101864-404
5-Acetyl-1-(2-chloro-3-pyridinyl)-2,4(1H,3H)-pyrimidinedione ≥95%
5-Acetyl-1-(2-chloro-3-pyridinyl)-2,4(1H,3H)-pyrimidinedione

Hazard: Irritant

Formula: C₁₁H₈ClN₃O₃
Melting Pt: 224…226 °C
MDL Number: MFCD00794645
CAS Number: 338413-91-5

Order Now

Specification Test Results

Melting point 224-226°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR