4,5-Diphenyl-1H-imidazol-2-ylhydrosulfide ≥95%

Supplier: Matrix Scientific

044127-5G 044127-1G 044127-500MG
101864-280EA 135.7 USD
101864-280 101864-282 101864-284
4,5-Diphenyl-1H-imidazol-2-ylhydrosulfide ≥95%
4,5-Diphenyl-1H-imidazol-2-ylhydrosulfide

Hazard: Irritant

Formula: C₁₅H₁₂N₂S
Melting Pt: 305…309 °C
MDL Number: MFCD00005189
CAS Number: 2349-58-8

Order Now

Specification Test Results

Melting point 305-309°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR