[1-Methyl-5-(4-methylphenoxy)-3-phenyl-1H-pyrazol-4-yl]methanol ≥95%
Supplier: Matrix Scientific
044124-5G
044124-1G
044124-500MG
101864-262EA
921.54
USD
101864-262
101864-264
101864-266
[1-Methyl-5-(4-methylphenoxy)-3-phenyl-1H-pyrazol-4-yl]methanol ≥95%
[1-Methyl-5-(4-methylphenoxy)-3-phenyl-1H-pyrazol-4-yl]methanol
Hazard: Irritant
Formula:
C₁₈H₁₈N₂O₂ Melting Pt: 150…152 °C |
MDL Number:
MFCD00244906 CAS Number: 318234-01-4 |
Specification Test Results
Melting point | 150-152°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...